ENA447957657 Enamine

methyl 2-(5-cyanopyridin-2-yl)acetate

  • MDL number MFCD18255619
  • Cas Number : 1256793-78-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205744

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205744
Availability details for
Quantity
  • Availability
  • Add to Cart