ENA447957679 Enamine

2-hydroxy-2-[3-(trifluoromethyl)phenyl]acetic acid

  • MDL number MFCD00014339
  • Cas Number : 349-10-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207720

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207720
Availability details for
Quantity
  • Availability
  • Add to Cart