ENA447957681 Enamine

1-(2-fluorophenyl)cyclopentane-1-carbonitrile

  • MDL number MFCD00800612
  • Cas Number : 214262-89-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205750

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205750
Availability details for
Quantity
  • Availability
  • Add to Cart