ENA447957685 Enamine

1-(2,3-dihydro-1H-indol-1-yl)prop-2-en-1-one

  • MDL number MFCD12091097
  • Cas Number : 104902-94-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207741

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207741
Availability details for
Quantity
  • Availability
  • Add to Cart