ENA447957687 Enamine

N-benzyl-N-methylazetidin-3-amine

  • MDL number MFCD16871783
  • Cas Number : 1339397-77-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. -20
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207743

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207743
Availability details for
Quantity
  • Availability
  • Add to Cart