ENA447957689 Enamine

3-[3-(trifluoromethyl)phenyl]piperidine

  • MDL number MFCD11047439
  • Cas Number : 64321-45-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207745

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207745
Availability details for
Quantity
  • Availability
  • Add to Cart