ENA447957690 Enamine

1-(2,2-diethoxyethyl)-4-fluorobenzene

  • MDL number MFCD11554373
  • Cas Number : 1743-98-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207746

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207746
Availability details for
Quantity
  • Availability
  • Add to Cart