ENA447957694 Enamine

2-[2-(2,6-dichlorophenyl)-1,3-thiazol-4-yl]-2-methylpropanoic acid

  • MDL number MFCD26420074
  • Cas Number : 1803612-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207754

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207754
Availability details for
Quantity
  • Availability
  • Add to Cart