ENA447957698 Enamine

2-amino-1-(4-methylphenyl)propan-1-ol

  • MDL number MFCD09035926
  • Cas Number : 176375-11-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207791

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207791
Availability details for
Quantity
  • Availability
  • Add to Cart