ENA447957700 Enamine

2-(4-nitrophenyl)propan-2-ol

  • MDL number MFCD00466951
  • Cas Number : 22357-57-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207793

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207793
Availability details for
Quantity
  • Availability
  • Add to Cart