ENA447957705 Enamine

1-(chloromethyl)-4-(2-methylbutan-2-yl)benzene

  • MDL number MFCD00018884
  • Cas Number : 28162-11-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207803

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207803
Availability details for
Quantity
  • Availability
  • Add to Cart