ENA447957706 Enamine

2-[4-(2-methoxyethanesulfonyl)phenyl]acetic acid

  • MDL number MFCD12411184
  • Cas Number : 1155610-14-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207804
Availability details for
Quantity
  • Availability
  • Add to Cart