ENA447957707 Enamine

1-(1,2,3,4-tetrahydroquinolin-1-yl)prop-2-en-1-one

  • MDL number MFCD12091081
  • Cas Number : 88611-48-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207807

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207807
Availability details for
Quantity
  • Availability
  • Add to Cart