ENA447957710 Enamine

1-(5-bromo-4-methylthiophen-2-yl)ethan-1-one

  • MDL number MFCD11110667
  • Cas Number : 859199-06-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207810

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207810
Availability details for
Quantity
  • Availability
  • Add to Cart