ENA447957712 Enamine

1-(3,4-dihydro-2H-1,4-benzoxazin-4-yl)prop-2-en-1-one

  • MDL number MFCD12091490
  • Cas Number : 1156925-29-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207812

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207812
Availability details for
Quantity
  • Availability
  • Add to Cart