ENA447957713 Enamine

2-(3-iodopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

  • MDL number MFCD26516403
  • Cas Number : 1422039-20-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207813
Availability details for
Quantity
  • Availability
  • Add to Cart