ENA447957714 Enamine

methyl 2-(piperazin-1-yl)butanoate

  • MDL number MFCD12818041
  • Cas Number : 1218333-65-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207815

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207815
Availability details for
Quantity
  • Availability
  • Add to Cart