ENA447957717 Enamine

3-(hydroxymethyl)-1lambda6-thiane-1,1-dione

  • MDL number MFCD17078852
  • Cas Number : 1243250-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205775

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205775
Availability details for
Quantity
  • Availability
  • Add to Cart