ENA447957719 Enamine

3-hydroxy-2-(4-methylphenyl)prop-2-enenitrile

  • Cas Number : 72899-87-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207824

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207824
Availability details for
Quantity
  • Availability
  • Add to Cart