ENA447957720 Enamine

methyl({[3-(thiophen-2-yl)-1,2,4-oxadiazol-5-yl]methyl})amine hydrochloride

  • MDL number MFCD11843649
  • Cas Number : 1185365-43-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205776

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205776
Availability details for
Quantity
  • Availability
  • Add to Cart