ENA447957723 Enamine

N,8-dimethyl-8-azabicyclo[3.2.1]octan-3-amine

  • MDL number MFCD19285176
  • Cas Number : 102312-28-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207833

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207833
Availability details for
Quantity
  • Availability
  • Add to Cart