ENA447957724 Enamine

methyl 5-bromo-6-hydroxy-2-methylpyridine-3-carboxylate

  • MDL number MFCD20232419
  • Cas Number : 1334499-25-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207840

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207840
Availability details for
Quantity
  • Availability
  • Add to Cart