ENA447957725 Enamine

5-bromo-6-hydroxy-2-methylpyridine-3-carboxylic acid

  • MDL number MFCD26938565
  • Cas Number : 1803604-56-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207841

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207841
Availability details for
Quantity
  • Availability
  • Add to Cart