ENA447957727 Enamine

1-(1-phenyl-1H-imidazol-2-yl)ethan-1-ol

  • MDL number MFCD28397493
  • Cas Number : 1508045-23-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207848

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207848
Availability details for
Quantity
  • Availability
  • Add to Cart