ENA447957729 Enamine

2-(4-ethylphenyl)-2-[(trimethylsilyl)oxy]acetonitrile

  • MDL number MFCD16786523
  • Cas Number : 1073135-75-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207864

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207864
Availability details for
Quantity
  • Availability
  • Add to Cart