ENA447957735 Enamine

N-[2-(aminomethyl)cyclohexyl]methanesulfonamide hydrochloride

  • MDL number MFCD28397498
  • Cas Number : 1591568-44-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207877

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207877
Availability details for
Quantity
  • Availability
  • Add to Cart