ENA447957737 Enamine

5,5-dimethylhexan-2-one

  • MDL number MFCD11553439
  • Cas Number : 14272-73-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205785
Availability details for
Quantity
  • Availability
  • Add to Cart