ENA447957739 Enamine

4-(4-tert-butylphenyl)cyclohexan-1-amine

  • MDL number MFCD21889587
  • Cas Number : 1369011-12-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205786
Availability details for
Quantity
  • Availability
  • Add to Cart