ENA447957744 Enamine

2-benzyl-2,7-diazaspiro[4.4]nonane

  • MDL number MFCD04115133
  • Cas Number : 885275-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207892

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207892
Availability details for
Quantity
  • Availability
  • Add to Cart