ENA447957748 Enamine

[2-(piperidin-1-yl)ethyl](propyl)amine

  • MDL number MFCD14646255
  • Cas Number : 99178-30-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207919
Availability details for
Quantity
  • Availability
  • Add to Cart