ENA447957755 Enamine

ethyl 2-bromo-5-(chlorosulfonyl)-4-fluorobenzoate

  • MDL number MFCD12169522
  • Cas Number : 1155084-72-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207927

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207927
Availability details for
Quantity
  • Availability
  • Add to Cart