ENA447957759 Enamine

5-chloro-2-methoxy-N-methyl-3-sulfamoylbenzamide

  • MDL number MFCD21225369
  • Cas Number : 1486433-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207933

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207933
Availability details for
Quantity
  • Availability
  • Add to Cart