ENA447957765 Enamine

[3-(naphthalen-2-yloxy)phenyl]methanamine

  • MDL number MFCD19551853
  • Cas Number : 1185096-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207942

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207942
Availability details for
Quantity
  • Availability
  • Add to Cart