ENA447957766 Enamine

3-(4-bromophenyl)aniline

  • MDL number MFCD11574739
  • Cas Number : 40641-71-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207943

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207943
Availability details for
Quantity
  • Availability
  • Add to Cart