ENA447957771 Enamine

2-chloro-5-formylbenzene-1-sulfonamide

  • MDL number MFCD18205751
  • Cas Number : 3279-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 167-170
Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207952
Availability details for
Quantity
  • Availability
  • Add to Cart