ENA447957772 Enamine

(2S,4R)-2-methyl-3,4-dihydro-2H-1-benzopyran-4-ol

  • MDL number MFCD31566974
  • Cas Number : 1820570-01-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207962

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207962
Availability details for
Quantity
  • Availability
  • Add to Cart