ENA447957774 Enamine

[6-(benzyloxy)pyridin-3-yl]methanol

  • MDL number MFCD14606379
  • Cas Number : 936344-83-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205814

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205814
Availability details for
Quantity
  • Availability
  • Add to Cart