ENA447957780 Enamine

4-(1-methyl-1H-pyrazol-5-yl)-1,2,3,6-tetrahydropyridine

  • MDL number MFCD11899301
  • Cas Number : 1215953-08-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Ice Pack
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-207978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-207978
Availability details for
Quantity
  • Availability
  • Add to Cart