ENA447957793 Enamine

3-[(4-fluorophenoxy)methyl]aniline

  • MDL number MFCD09803139
  • Cas Number : 1016531-20-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208049

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208049
Availability details for
Quantity
  • Availability
  • Add to Cart