ENA447957796 Enamine

ethyl 3-methyl-4-oxopent-2-enoate

  • Cas Number : 107368-27-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205860

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205860
Availability details for
Quantity
  • Availability
  • Add to Cart