ENA447957797 Enamine

(2,6-dichloro-3-fluorophenyl)methanol

  • MDL number MFCD16660033
  • Cas Number : 1227611-90-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208054

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208054
Availability details for
Quantity
  • Availability
  • Add to Cart