ENA447957798 Enamine

4-methylthiolan-3-amine

  • MDL number MFCD19212770
  • Cas Number : 115660-79-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208055
Availability details for
Quantity
  • Availability
  • Add to Cart