ENA447957799 Enamine

1-(azepan-2-yl)propan-2-ol

  • MDL number MFCD21938930
  • Cas Number : 1506449-63-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208056

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208056
Availability details for
Quantity
  • Availability
  • Add to Cart