ENA447957804 Enamine

3-amino-3-(3,4-dichlorophenyl)propan-1-ol

  • MDL number MFCD03412661
  • Cas Number : 147611-60-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208069

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208069
Availability details for
Quantity
  • Availability
  • Add to Cart