ENA447957807 Enamine

3-methyl-1,3,7-triazaspiro[4.4]nonane-2,4-dione hydrochloride

  • MDL number MFCD26792909
  • Cas Number : 28937-15-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208078

Safety Information

signalword Warning
pcodes P312
MSDS https://www.enaminestore.com/catalog/EN300-208078
hstatements H303,H313
Availability details for
Quantity
  • Availability
  • Add to Cart