ENA447957811 Enamine

2-methyl-2-(2-methylpropyl)-1,3-dioxolane

  • MDL number MFCD22055414

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208108
Availability details for
Quantity
  • Availability
  • Add to Cart