ENA447957825 Enamine

1-[(4-methoxyphenyl)methyl]-1H-pyrazol-4-amine

  • MDL number MFCD11128222
  • Cas Number : 1152867-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205908

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205908
Availability details for
Quantity
  • Availability
  • Add to Cart