ENA447957839 Enamine

3-[3-(3-hydroxypropyl)phenyl]propan-1-ol

  • MDL number MFCD09926774
  • Cas Number : 41009-85-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208174
Availability details for
Quantity
  • Availability
  • Add to Cart