ENA447957840 Enamine

3-amino-2-cyclobutylpropan-1-ol

  • MDL number MFCD28122738
  • Cas Number : 1706444-88-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-208176

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208176
Availability details for
Quantity
  • Availability
  • Add to Cart