ENA447957842 Enamine

6-[(methylamino)methyl]-3,4-dihydro-2H-1,4-benzoxazin-3-one

  • MDL number MFCD19375025
  • Cas Number : 1267218-13-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-205915

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-205915
Availability details for
Quantity
  • Availability
  • Add to Cart